accessibility menu, dialog, popup

UserName

CAS RN 652145-28-3

NONSSOOH(E/Z)

CAS RN 652145-28-3

IUPAC Name: 2-[5-({4-[4-(2,2-diphenylethenyl)phenyl]-1H,2H,3H,3aH,4H,8bH-cyclopenta[b]indol-7-yl}methylidene)-4-oxo-2-sulfanylidene-1,3-thiazolidin-3-yl]acetic acid
Synonyms: [5-({4-[4-(2,2-diphenylethenyl)phenyl]-1H,2H,3H,3aH,8bH-cyclopenta[b]indol-7-yl}methylidene)-4-oxo-2-sulfanylidene-1,3-thiazolidin-3-yl]acetic acid
Molecular Weight (g/mol): 614.78
Molecular Formula: C37H30N2O3S2
InChi Key: XGMCROHUTRXETK-UHFFFAOYNA-N
SMILES: OC(=O)CN1C(=S)SC(=CC2=CC3=C(C=C2)N(C2CCCC32)C2=CC=C(C=C(C3=CC=CC=C3)C3=CC=CC=C3)C=C2)C1=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results