accessibility menu, dialog, popup

UserName

H3CS

CAS RN 65016-55-9

IUPAC Name: 3-decylthiophene
Synonyms: 3-decylthiophene
Molecular Weight (g/mol): 224.41
Molecular Formula: C14H24S
InChi Key: JAYBIBLZTQMCAY-UHFFFAOYSA-N
SMILES: CCCCCCCCCCC1=CSC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results