CAS RN 647-12-1
CAS RN 647-12-1
IUPAC Name:
pentadecafluorooctanenitrile
Synonyms:
pentadecafluorooctanenitrile
Molecular Weight (g/mol):
395.07
Molecular Formula:
C8F15N
InChi Key:
FULLNJNBFUCVES-UHFFFAOYSA-N
SMILES:
FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C#N