accessibility menu, dialog, popup

UserName

CAS RN 64394-47-4

IUPAC Name: 5-{5-oxo-2H,5H-[1,3]dithiolo[4,5-d][1,3]dithiol-2-ylidene}-2H,5H-[1,3]dithiolo[4,5-d][1,3]dithiol-2-one
Synonyms: 5-{5-oxo-[1,3]dithiolo[4,5-d][1,3]dithiol-2-ylidene}-[1,3]dithiolo[4,5-d][1,3]dithiol-2-one
Molecular Weight (g/mol): 384.57
Molecular Formula: C8O2S8
InChi Key: JVSSQMMIVZRMMO-UHFFFAOYSA-N
SMILES: O=C1SC2=C(S1)SC(S2)=C1SC2=C(SC(=O)S2)S1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results