CAS RN 64339-95-3
IUPAC Name:
(3R,4R)-4-(benzoyloxy)-2,5-dioxooxolan-3-yl benzoate
Synonyms:
(3R,4R)-4-(benzoyloxy)-2,5-dioxooxolan-3-yl benzoate
Molecular Weight (g/mol):
340.29
Molecular Formula:
C18H12O7
InChi Key:
OXIKRMSPXYQFOT-ZCWZLOQUNA-N
SMILES:
O=C(O[C@@H]1[C@@H](OC(=O)C2=CC=CC=C2)C(=O)OC1=O)C1=CC=CC=C1
1
–
1
of
1
results
1
Promotions Available
(+)-Dibenzoyl-L-tartaric anhydride, 98%
(+)-Dibenzoyl-L-tartaric anhydride, 98%
CAS: 64339-95-3 Molecular Formula: C18H12O7 Molecular Weight (g/mol): 340.29 MDL Number: MFCD00067102 InChI Key: OXIKRMSPXYQFOT-ZCWZLOQUNA-N Synonym: 3r,4r-2,5-dioxotetrahydrofuran-3,4-diyl dibenzoate,+-dibenzoyl-l-tartaric anhydride,2,5-furandione, 3,4-bis benzoyloxy dihydro-, 3r,4r,2-o,3-o-dibenzoyl-l-tartaric anhydride,3r,4r-2,5-dioxo-3,4-dibenzoyloxy-3,4-dihydrofuran,3r,4r-4-benzoyloxy-2,5-dioxooxolan-3-yl benzoate PubChem CID: 2733743 IUPAC Name: [(3R,4R)-4-benzoyloxy-2,5-dioxooxolan-3-yl] benzoate SMILES: O=C(O[C@@H]1[C@@H](OC(=O)C2=CC=CC=C2)C(=O)OC1=O)C1=CC=CC=C1