accessibility menu, dialog, popup

UserName

CAS RN 64328-68-3

IUPAC Name: 2-bromo-1-(4-pentylphenyl)ethan-1-one
Synonyms: 2-bromo-1-(4-pentylphenyl)ethanone
Molecular Weight (g/mol): 269.18
Molecular Formula: C13H17BrO
InChi Key: AVYNDJWQMUOSJZ-UHFFFAOYSA-N
SMILES: CCCCCC1=CC=C(C=C1)C(=O)CBr

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)