CAS RN 642995-16-2
IUPAC Name:
2-{1,3-dioxo-1H,2H,3H-benzo[f]isoindol-2-yl}cyclohexane-1-carboxylic acid
Synonyms:
2-{1,3-dioxobenzo[f]isoindol-2-yl}cyclohexane-1-carboxylic acid
Molecular Weight (g/mol):
323.35
Molecular Formula:
C19H17NO4
InChi Key:
RPVXICJCKYVHHZ-UHFFFAOYNA-N
SMILES:
OC(=O)C1CCCCC1N1C(=O)C2=C(C=C3C=CC=CC3=C2)C1=O
1
–
1
of
1
results
1
(1S,2S)-2-(Naphthalene-2,3-dicarboximido)cyclohexanecarboxylic Acid 98.0+%, TCI America™
CAS: 642995-16-2 Molecular Formula: C19H17NO4 Molecular Weight (g/mol): 323.348 InChI Key: RPVXICJCKYVHHZ-BBRMVZONSA-N Synonym: N-[(1S,2S)-2-Carboxycyclohexyl]naphthalene-2,3-dicarboximide PubChem CID: 44629757 IUPAC Name: (1S,2S)-2-(1,3-dioxobenzo[f]isoindol-2-yl)cyclohexane-1-carboxylic acid SMILES: C1CCC(C(C1)C(=O)O)N2C(=O)C3=CC4=CC=CC=C4C=C3C2=O