accessibility menu, dialog, popup

UserName

NaH3CCH3NSOOOH3CH3CNSOOO(E)(E)(E/Z)

CAS RN 64285-36-5

IUPAC Name: sodium 2-[(3E)-5-[(2E)-1,1-dimethyl-3-(4-sulfonatobutyl)-1H,2H,3H-benzo[e]indol-2-ylidene]penta-1,3-dien-1-yl]-1,1-dimethyl-3-(4-sulfonatobutyl)-1H-benzo[e]indol-3-ium
Synonyms: sodium 2-[(3E)-5-[(2E)-1,1-dimethyl-3-(4-sulfonatobutyl)benzo[e]indol-2-ylidene]penta-1,3-dien-1-yl]-1,1-dimethyl-3-(4-sulfonatobutyl)benzo[e]indol-3-ium
Molecular Weight (g/mol): 748.93
Molecular Formula: C41H45N2NaO6S2
InChi Key: LAYIHRUYRMYLTJ-UHFFFAOYSA-M
SMILES: [Na+].CC1(C)\C(=C/C=C/C=CC2=[N+](CCCCS([O-])(=O)=O)C3=CC=C4C=CC=CC4=C3C2(C)C)N(CCCCS([O-])(=O)=O)C2=CC=C3C=CC=CC3=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results