accessibility menu, dialog, popup

UserName

H3COONH3COONH3COONCH3

CAS RN 64265-57-2

IUPAC Name: 2,2-bis({[3-(2-methylaziridin-1-yl)propanoyl]oxy}methyl)butyl 3-(2-methylaziridin-1-yl)propanoate
Synonyms: 2,2-bis({[3-(2-methylaziridin-1-yl)propanoyl]oxy}methyl)butyl 3-(2-methylaziridin-1-yl)propanoate
Molecular Weight (g/mol): 467.61
Molecular Formula: C24H41N3O6
InChi Key: YKEGOEUSKXVSPN-UHFFFAOYNA-N
SMILES: CCC(COC(=O)CCN1CC1C)(COC(=O)CCN1CC1C)COC(=O)CCN1CC1C
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results