CAS RN 6421-30-3
CAS RN 6421-30-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Ethyl 4-[(4-Methoxybenzylidene)amino]cinnamate 98.0+%, TCI Americaâ„¢
CAS: 6421-30-3 Molecular Formula: C19H19NO3 Molecular Weight (g/mol): 309.365 MDL Number: MFCD00059231 InChI Key: FMNNRRPDXZWUAE-KGCZTCBMSA-N Synonym: 4-(p-Anisalamino)cinnamic Acid Ethyl Ester, 4-[(p-Anisylidene)amino]cinnamic Acid Ethyl Ester, Ethyl 4-(p-Anisalamino)cinnamate, Ethyl 4-[(p-Anisylidene)amino]cinnamate, 4-[(4-Methoxybenzylidene)amino]cinnamic Acid Ethyl Ester PubChem CID: 780892 IUPAC Name: ethyl (E)-3-[4-[(4-methoxyphenyl)methylideneamino]phenyl]prop-2-enoate SMILES: CCOC(=O)C=CC1=CC=C(C=C1)N=CC2=CC=C(C=C2)OC