accessibility menu, dialog, popup

UserName

H3CO

CAS RN 638-66-4

IUPAC Name: octadecanal
Synonyms: octadecanal
Molecular Weight (g/mol): 268.49
Molecular Formula: C18H36O
InChi Key: FWWQKRXKHIRPJY-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCC=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results