accessibility menu, dialog, popup

UserName

H2NONHOHNONHOHO

CAS RN 637-84-3

IUPAC Name: 2-{2-[2-(2-aminoacetamido)acetamido]acetamido}acetic acid
Synonyms: {2-[2-(2-aminoacetamido)acetamido]acetamido}acetic acid
Molecular Weight (g/mol): 246.22
Molecular Formula: C8H14N4O5
InChi Key: QMOQBVOBWVNSNO-UHFFFAOYSA-N
SMILES: NCC(=O)NCC(=O)NCC(=O)NCC(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results