accessibility menu, dialog, popup

UserName

ClHNOO

CAS RN 6344-05-4

IUPAC Name: 4-chloro-2,3-dihydro-1H-indole-2,3-dione
Synonyms: 4-chloro-1H-indole-2,3-dione
Molecular Weight (g/mol): 181.58
Molecular Formula: C8H4ClNO2
InChi Key: HSYFISNDMZKGRS-UHFFFAOYSA-N
SMILES: ClC1=CC=CC2=C1C(=O)C(=O)N2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results