accessibility menu, dialog, popup

UserName

HOOOBr

CAS RN 6340-79-0

IUPAC Name: 4-(4-bromophenyl)-4-oxobutanoic acid
Synonyms: 4-(4-bromophenyl)-4-oxobutanoic acid
Molecular Weight (g/mol): 257.08
Molecular Formula: C10H9BrO3
InChi Key: ZODFRCZNTXLDDW-UHFFFAOYSA-N
SMILES: OC(=O)CCC(=O)C1=CC=C(Br)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results