accessibility menu, dialog, popup

UserName

H3CHNOONH

CAS RN 6309-61-1

IUPAC Name: 6-methyl-1,2,3,4-tetrahydroquinoxaline-2,3-dione
Synonyms: 6-methyl-1,4-dihydroquinoxaline-2,3-dione
Molecular Weight (g/mol): 176.18
Molecular Formula: C9H8N2O2
InChi Key: HOHZZPCLZWZMOM-UHFFFAOYSA-N
SMILES: CC1=CC=C2NC(=O)C(=O)NC2=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results