CAS RN 63-64-9
IUPAC Name:
[(3,4-dimethoxyphenyl)methyl](methyl)azanium
Synonyms:
[(3,4-dimethoxyphenyl)methyl](methyl)azanium
Molecular Weight (g/mol):
182.24
Molecular Formula:
C10H16NO2
InChi Key:
XQODFBIAQVJQHF-UHFFFAOYSA-O
SMILES:
C[NH2+]CC1=CC=C(OC)C(OC)=C1
1
–
1
of
1
results
1
Promotions Available
3,4-Dimethoxy-N-methylbenzylamine, 97%, Thermo Scientific™
3,4-Dimethoxy-N-methylbenzylamine, 97%, Thermo Scientific™
CAS: 63-64-9 Molecular Formula: C10H15NO2 Molecular Weight (g/mol): 181.24 InChI Key: XQODFBIAQVJQHF-UHFFFAOYSA-N Synonym: 3,4-dimethoxybenzyl methylamine,n-methyl-3,4-dimethoxybenzylamine,1-3,4-dimethoxyphenyl-n-methylmethanamine,3,4-dimethoxyphenyl methyl methyl amine,3,4-dimethoxy-n-methylbenzylamine,3,4-dimethoxy-benzyl-methyl-amine,benzenemethanamine, 3,4-dimethoxy-n-methyl,1-3,4-dimethoxyphenyl-n-methyl-methanamine,3,4-dimethoxyphenyl methyl methylamine,3,4-dimethoxybenzyl-n-methylamine PubChem CID: 592161 IUPAC Name: 1-(3,4-dimethoxyphenyl)-N-methylmethanamine SMILES: CNCC1=CC(=C(C=C1)OC)OC