CAS RN 6294-79-7
IUPAC Name:
1,3-bis(prop-2-en-1-yl)-1,3,5-triazinane-2,4,6-trione
Synonyms:
1,3-bis(prop-2-en-1-yl)-1,3,5-triazinane-2,4,6-trione
Molecular Weight (g/mol):
209.21
Molecular Formula:
C9H11N3O3
InChi Key:
UCBVELLBUAKUNE-UHFFFAOYSA-N
SMILES:
C=CCN1C(=O)NC(=O)N(CC=C)C1=O
1
–
1
of
1
results
1
Diallyl Isocyanurate 98.0+%, TCI America™
CAS: 6294-79-7 Molecular Formula: C9H11N3O3 Molecular Weight (g/mol): 209.205 MDL Number: MFCD00059763 InChI Key: UCBVELLBUAKUNE-UHFFFAOYSA-N Synonym: Isocyanuric Acid Diallyl Ester PubChem CID: 94974 IUPAC Name: 1,3-bis(prop-2-enyl)-1,3,5-triazinane-2,4,6-trione SMILES: C=CCN1C(=O)NC(=O)N(C1=O)CC=C