accessibility menu, dialog, popup

UserName

CAS RN 6294-40-2

IUPAC Name: 1-bromo-4-methylcyclohexane
Synonyms: 1-bromo-4-methylcyclohexane
Molecular Weight (g/mol): 177.09
Molecular Formula: C7H13Br
InChi Key: RLOHLPHAOHGRNM-UHFFFAOYSA-N
SMILES: CC1CCC(Br)CC1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)