accessibility menu, dialog, popup

UserName

H3CSOONNH

CAS RN 62937-96-6

IUPAC Name: 1-(ethanesulfonyl)piperazine
Synonyms: 1-(ethanesulfonyl)piperazine
Molecular Weight (g/mol): 178.25
Molecular Formula: C6H14N2O2S
InChi Key: BIYGAOBOLDXNHM-UHFFFAOYSA-N
SMILES: CCS(=O)(=O)N1CCNCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results