CAS RN 6287-83-8
IUPAC Name:
1H-1,3-benzodiazole-6-carbonitrile
Synonyms:
3H-1,3-benzodiazole-5-carbonitrile
Molecular Weight (g/mol):
143.15
Molecular Formula:
C8H5N3
InChi Key:
NICYTXJGZRYCEQ-UHFFFAOYSA-N
SMILES:
N#CC1=CC=C2N=CNC2=C1
1
–
1
of
1
results
1
Promotions Available
Benzimidazole-6-carbonitrile, 97%, Thermo Scientific Chemicals
Benzimidazole-6-carbonitrile, 97%, Thermo Scientific Chemicals
CAS: 6287-83-8 Molecular Formula: C8H5N3 Molecular Weight (g/mol): 143.15 MDL Number: MFCD06809988 InChI Key: NICYTXJGZRYCEQ-UHFFFAOYSA-N Synonym: 1h-benzimidazole-5-carbonitrile,1h-benzo d imidazole-6-carbonitrile,1h-1,3-benzodiazole-5-carbonitrile,1h-benzo d imidazole-5-carbonitrile,5-cyanobenzimidazole,1h-benzimidazole-6-carbonitrile,5-benzimidazolecarbonitrile,benzimidazole-5-carbonitrile,1h-benzoimidazole-5-carbonitrile,3h-1,3-benzodiazole-5-carbonitrile PubChem CID: 223906 IUPAC Name: 1H-1,3-benzodiazole-6-carbonitrile SMILES: N#CC1=CC=C2N=CNC2=C1