accessibility menu, dialog, popup

UserName

CAS RN 628-11-5

ClOClO

CAS RN 628-11-5

IUPAC Name: 3-chloropropyl carbonochloridate
Synonyms: 3-chloropropyl carbonochloridate
Molecular Weight (g/mol): 156.99
Molecular Formula: C4H6Cl2O2
InChi Key: MTXMEFUEBCFWCY-UHFFFAOYSA-N
SMILES: ClCCCOC(Cl)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results