accessibility menu, dialog, popup

UserName

H3COOCl

CAS RN 628-09-1

IUPAC Name: 3-chloropropyl acetate
Synonyms: 1-propanol, 3-chloro-, acetate
Molecular Weight (g/mol): 136.58
Molecular Formula: C5H9ClO2
InChi Key: KPOHQIPNNIMWRL-UHFFFAOYSA-N
SMILES: CC(=O)OCCCCl
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)