accessibility menu, dialog, popup

UserName

NaNaCH3HNCH3NH3CH3CSHOOOSOOOH3CNCH3NNSOOO(E)(E/Z)(E/Z)

CAS RN 62758-15-0

IUPAC Name: disodium 4-{2-[4-(dimethylamino)phenyl]diazen-1-yl}benzene-1-sulfonate 4-{[4-(ethylamino)-3-methylphenyl][(1E)-4-(ethylimino)-3-methylcyclohexa-2,5-dien-1-ylidene]methyl}-3-sulfobenzene-1-sulfonate
Synonyms: disodium methyl orange 4-{[4-(ethylamino)-3-methylphenyl][(1E)-4-(ethylimino)-3-methylcyclohexa-2,5-dien-1-ylidene]methyl}-3-sulfobenzenesulfonate
Molecular Weight (g/mol): 865.94
Molecular Formula: C39H41N5Na2O9S3
InChi Key: QZATZDKHJDOZFD-DGVIJJPGSA-L
SMILES: [Na+].[Na+].CN(C)C1=CC=C(C=C1)N=NC1=CC=C(C=C1)S([O-])(=O)=O.CCNC1=CC=C(C=C1C)C(=C1\C=CC(=NCC)C(C)=C1)\C1=CC=C(C=C1S(O)(=O)=O)S([O-])(=O)=O
Color 
  • (1)
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)