accessibility menu, dialog, popup

UserName

CH3OOCH3OOCH3OOEu

CAS RN 62667-64-5

IUPAC Name: europium(3+) triacetate
Synonyms: europium(3+) triacetate
Molecular Weight (g/mol): 329.1
Molecular Formula: C6H9EuO6
InChi Key: LNYNHRRKSYMFHF-UHFFFAOYSA-K
SMILES: [Eu+3].CC([O-])=O.CC([O-])=O.CC([O-])=O
Grade 
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results