CAS RN 626232-27-7
IUPAC Name:
potassium trimethoxy(trifluoromethyl)boranuide
Synonyms:
potassium trimethoxy(trifluoromethyl)boranuide
Molecular Weight (g/mol):
212.02
Molecular Formula:
C4H9BF3KO3
InChi Key:
XQUUVPNLRZQWNF-UHFFFAOYSA-N
SMILES:
[K+].CO[B-](OC)(OC)C(F)(F)F
1
–
1
of
1
results
1
Potassium Trimethoxy(trifluoromethyl)borate 95.0+%, TCI America™
CAS: 626232-27-7 Molecular Formula: C4H9BF3KO3 Molecular Weight (g/mol): 212.017 MDL Number: MFCD22124929 InChI Key: XQUUVPNLRZQWNF-UHFFFAOYSA-N Synonym: Potassium (Trifluoromethyl)trimethoxyborate PubChem CID: 23682739 IUPAC Name: potassium;trimethoxy(trifluoromethyl)boranuide SMILES: [B-](C(F)(F)F)(OC)(OC)OC.[K+]