CAS RN 625-36-5
IUPAC Name:
3-chloropropanoyl chloride
Synonyms:
propanoyl chloride, 3-chloro-
Molecular Weight (g/mol):
126.96
Molecular Formula:
C3H4Cl2O
InChi Key:
INUNLMUAPJVRME-UHFFFAOYSA-N
SMILES:
ClCCC(Cl)=O
1
–
1
of
1
results
1
Promotions Available
3-Chloropropionyl chloride, 97%
3-Chloropropionyl chloride, 97%
CAS: 625-36-5 Molecular Formula: C3H4Cl2O Molecular Weight (g/mol): 126.96 MDL Number: MFCD00000747 InChI Key: INUNLMUAPJVRME-UHFFFAOYSA-N Synonym: 3-chloropropionyl chloride,propanoyl chloride, 3-chloro,propionyl chloride, 3-chloro,3-chloropropionylchloride,beta-chloropropionyl chloride,3-chloropropionic acid chloride,chloropropionyl chloride,propanoylchloride, 3-chloro,.beta.-chloropropanoyl chloride,.beta.-chloropropionyl chloride PubChem CID: 69364 IUPAC Name: 3-chloropropanoyl chloride SMILES: ClCCC(Cl)=O