accessibility menu, dialog, popup

UserName

CAS RN 6237-59-8

CH3OOOOCH3OOCH3OOCH3OOH3COOH3C

CAS RN 6237-59-8

IUPAC Name: 1,2,3,4,5,6-hexamethyl benzene-1,2,3,4,5,6-hexacarboxylate
Synonyms: 1,2,3,4,5,6-hexamethyl benzene-1,2,3,4,5,6-hexacarboxylate
Molecular Weight (g/mol): 426.33
Molecular Formula: C18H18O12
InChi Key: BQLICNRRYLYEDI-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(C(=O)OC)C(C(=O)OC)=C(C(=O)OC)C(C(=O)OC)=C1C(=O)OC
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results