accessibility menu, dialog, popup

UserName

CAS RN 623-56-3

H3COCH3CH3

CAS RN 623-56-3

IUPAC Name: 5-methylhexan-3-one
Synonyms: 5-methyl-3-hexanone
Molecular Weight (g/mol): 114.19
Molecular Formula: C7H14O
InChi Key: DXVYLFHTJZWTRF-UHFFFAOYSA-N
SMILES: CCC(=O)CC(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results