accessibility menu, dialog, popup

UserName

CAS RN 623-30-3

OO(E)

CAS RN 623-30-3

IUPAC Name: (2E)-3-(furan-2-yl)prop-2-enal
Synonyms: 3-(2-furyl)acrylaldehyde
Molecular Weight (g/mol): 122.12
Molecular Formula: C7H6O2
InChi Key: VZIRCHXYMBFNFD-HNQUOIGGSA-N
SMILES: O=C\C=C\C1=CC=CO1
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)