accessibility menu, dialog, popup

UserName

H3CCH3NH2

CAS RN 62281-06-5

IUPAC Name: 9-(aminomethyl)nonadecane
Synonyms: 9-(aminomethyl)nonadecane
Molecular Weight (g/mol): 297.57
Molecular Formula: C20H43N
InChi Key: VDNQHHBRKZQBPY-UHFFFAOYNA-N
SMILES: CCCCCCCCCCC(CN)CCCCCCCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results