accessibility menu, dialog, popup

UserName

CAS RN 62224-24-2

IUPAC Name: methyl 4,5-dibromothiophene-2-carboxylate
Synonyms: methyl 4,5-dibromothiophene-2-carboxylate
Molecular Weight (g/mol): 299.96
Molecular Formula: C6H4Br2O2S
InChi Key: URPMSPAEAVFKCO-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(Br)=C(Br)S1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results