accessibility menu, dialog, popup

UserName

CAS RN 622-39-9

IUPAC Name: 2-propylpyridine
Synonyms: pyridine, 2-propyl-
Molecular Weight (g/mol): 121.18
Molecular Formula: C8H11N
InChi Key: OIALIKXMLIAOSN-UHFFFAOYSA-N
SMILES: CCCC1=CC=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results