accessibility menu, dialog, popup

UserName

CAS RN 6206-25-3

IUPAC Name: methyl 1-methylcyclopropane-1-carboxylate
Synonyms: methyl 1-methylcyclopropane-1-carboxylate
Molecular Weight (g/mol): 114.14
Molecular Formula: C6H10O2
InChi Key: COHFTBWCEOBGOR-UHFFFAOYSA-N
SMILES: COC(=O)C1(C)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results