accessibility menu, dialog, popup

UserName

H3CCl

CAS RN 62016-82-4

IUPAC Name: 1-chlorotriacontane
Synonyms: 1-chlorotriacontane
Molecular Weight (g/mol): 457.27
Molecular Formula: C30H61Cl
InChi Key: NSPRLRIWTSAZOP-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCl
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results