accessibility menu, dialog, popup

UserName

N

CAS RN 620-95-1

IUPAC Name: 3-benzylpyridine
Synonyms: 3-benzylpyridine
Molecular Weight (g/mol): 169.23
Molecular Formula: C12H11N
InChi Key: UUCLVSDUMKMBSM-UHFFFAOYSA-N
SMILES: C(C1=CC=CC=C1)C1=CC=CN=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results