accessibility menu, dialog, popup

UserName

NaH3COSOOO

CAS RN 61827-67-6

IUPAC Name: sodium 4-acetylbenzene-1-sulfonate
Synonyms: sodium 4-acetylbenzenesulfonate
Molecular Weight (g/mol): 222.19
Molecular Formula: C8H7NaO4S
InChi Key: PUWCULQGLSPCMQ-UHFFFAOYSA-M
SMILES: [Na+].CC(=O)C1=CC=C(C=C1)S([O-])(=O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results