accessibility menu, dialog, popup

UserName

H3COCH3O

CAS RN 6175-23-1

IUPAC Name: nonane-2,4-dione
Synonyms: nonane-2,4-dione
Molecular Weight (g/mol): 156.23
Molecular Formula: C9H16O2
InChi Key: KFBXUKHERGLHLG-UHFFFAOYSA-N
SMILES: CCCCCC(=O)CC(C)=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)