accessibility menu, dialog, popup

UserName

H3CN

CAS RN 617-92-5

IUPAC Name: 1-ethyl-1H-pyrrole
Synonyms: 1H-pyrrole, 1-ethyl-
Molecular Weight (g/mol): 95.15
Molecular Formula: C6H9N
InChi Key: VPUAYOJTHRDUTK-UHFFFAOYSA-N
SMILES: CCN1C=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results