accessibility menu, dialog, popup

UserName

CAS RN 617-68-5

HClH2NNH2OHO

CAS RN 617-68-5

IUPAC Name: hydrogen 2,6-diaminohexanoic acid chloride
Synonyms: hydrogen lysine chloride
Molecular Weight (g/mol): 182.65
Molecular Formula: C6H15ClN2O2
InChi Key: BVHLGVCQOALMSV-UHFFFAOYNA-N
SMILES: [H+].[Cl-].NCCCCC(N)C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results