accessibility menu, dialog, popup

UserName

CAS RN 617-48-1

IUPAC Name: 2-hydroxybutanedioic acid
Synonyms: malic acid H2mal aepfelsaeure apple acid
Molecular Weight (g/mol): 134.09
Molecular Formula: C4H6O5
InChi Key: BJEPYKJPYRNKOW-UHFFFAOYNA-N
SMILES: OC(CC(O)=O)C(O)=O
Grade 
  • (4)
Percent Purity 
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (4)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)