accessibility menu, dialog, popup

UserName

CH3OOOO

CAS RN 61683-99-6

IUPAC Name: 5-(4-methyl-1,3-dioxolan-2-yl)-2H-1,3-benzodioxole
Synonyms: 5-(4-methyl-1,3-dioxolan-2-yl)-2H-1,3-benzodioxole
Molecular Weight (g/mol): 208.21
Molecular Formula: C11H12O4
InChi Key: HXNSADTWQWLFCB-UHFFFAOYNA-N
SMILES: CC1COC(O1)C1=CC=C2OCOC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results