accessibility menu, dialog, popup

UserName

CH3NH

CAS RN 616-43-3

IUPAC Name: 3-methyl-1H-pyrrole
Synonyms: 3-methylpyrrole
Molecular Weight (g/mol): 81.12
Molecular Formula: C5H7N
InChi Key: FEKWWZCCJDUWLY-UHFFFAOYSA-N
SMILES: CC1=CNC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results