accessibility menu, dialog, popup

UserName

H3COONCH3NO

CAS RN 615-53-2

IUPAC Name: N-methyl-N'-oxoethoxycarbohydrazide
Synonyms: ethyl N-methylnitrosocarbamate
Molecular Weight (g/mol): 132.12
Molecular Formula: C4H8N2O3
InChi Key: CAUBWLYZCDDYEF-UHFFFAOYSA-N
SMILES: CCOC(=O)N(C)N=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results