accessibility menu, dialog, popup

UserName

ONON

CAS RN 613-50-3

IUPAC Name: 6-nitroquinoline
Synonyms: 6-nitroquinoline
Molecular Weight (g/mol): 174.16
Molecular Formula: C9H6N2O2
InChi Key: SMHPLBXIVNQFBA-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C2N=CC=CC2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results