accessibility menu, dialog, popup

UserName

N

CAS RN 613-46-7

IUPAC Name: naphthalene-2-carbonitrile
Synonyms: 2-naphthalenecarbonitrile
Molecular Weight (g/mol): 153.18
Molecular Formula: C11H7N
InChi Key: AZKDTTQQTKDXLH-UHFFFAOYSA-N
SMILES: N#CC1=CC=C2C=CC=CC2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results