accessibility menu, dialog, popup

UserName

H3COH3CCH3OOOCH3OCH3OH3C(?)

CAS RN 61281-37-6

IUPAC Name: 3,4,5,19-tetramethoxy-9,10-dimethyl-15,17-dioxatetracyclo[10.7.0.0²,⁷.0¹⁴,¹⁸]nonadeca-1(19),2(7),3,5,12,14(18)-hexaene
Synonyms: 3,4,5,19-tetramethoxy-9,10-dimethyl-15,17-dioxatetracyclo[10.7.0.0²,⁷.0¹⁴,¹⁸]nonadeca-1(19),2(7),3,5,12,14(18)-hexaene
Molecular Weight (g/mol): 400.47
Molecular Formula: C23H28O6
InChi Key: RTZKSTLPRTWFEV-UHFFFAOYNA-N
SMILES: COC1=CC2=C(C(OC)=C1OC)C1=C(OC)C3=C(OCO3)C=C1CC(C)C(C)C2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results