CAS RN 61204-01-1
IUPAC Name:
4-(4-pentylcyclohexyl)benzonitrile
Synonyms:
4-(4-pentylcyclohexyl)benzonitrile
Molecular Weight (g/mol):
255.41
Molecular Formula:
C18H25N
InChi Key:
FURZYCFZFBYJBT-UHFFFAOYSA-N
SMILES:
CCCCCC1CCC(CC1)C1=CC=C(C=C1)C#N
1
–
1
of
1
results
1
4-(trans-4-Amylcyclohexyl)benzonitrile 98.0+%, TCI America™
CAS: 61204-01-1 Molecular Formula: C18H25N Molecular Weight (g/mol): 255.41 MDL Number: MFCD00075548 InChI Key: FURZYCFZFBYJBT-UHFFFAOYSA-N Synonym: trans-4-4-pentylcyclohexyl benzonitrile,4-trans-4'-n-pentylcyclohexyl benzonitrile,4-trans-4-pentylcyclohexyl benzonitrile,4-4-pentylcyclohexyl benzonitrile,4-trans-4-pentylcyclohexyl-benzonitrile,4-trans-4-n-pentylcyclohexyl benzonitrile,benzonitrile, 4-trans-4-pentylcyclohexyl,4-trans-4-amylcyclohexyl benzonitrile,benzonitrile, 4-4-pentylcyclohexyl,1-trans-4-amylcyclohexyl-4-cyanobenzene PubChem CID: 109063 IUPAC Name: 4-(4-pentylcyclohexyl)benzonitrile SMILES: CCCCCC1CCC(CC1)C1=CC=C(C=C1)C#N