accessibility menu, dialog, popup

UserName

CH3N

CAS RN 61203-99-4

IUPAC Name: 4-(4-propylcyclohexyl)benzonitrile
Synonyms: 4-(4-propylcyclohexyl)benzonitrile
Molecular Weight (g/mol): 227.35
Molecular Formula: C16H21N
InChi Key: XXUSEPRYHRDKFV-UHFFFAOYSA-N
SMILES: CCCC1CCC(CC1)C1=CC=C(C=C1)C#N
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results