accessibility menu, dialog, popup

UserName

CAS RN 60853-81-8

IUPAC Name: 2-(dimethylamino)acetyl chloride hydrochloride
Synonyms: 2-(dimethylamino)acetyl chloride hydrochloride
Molecular Weight (g/mol): 158.02
Molecular Formula: C4H9Cl2NO
InChi Key: MVAXDKDOPWPFML-UHFFFAOYSA-N
SMILES: Cl.CN(C)CC(Cl)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results