accessibility menu, dialog, popup

UserName

ON

CAS RN 607-00-1

IUPAC Name: N,N-diphenylformamide
Synonyms: formamide, N,N-diphenyl-
Molecular Weight (g/mol): 197.24
Molecular Formula: C13H11NO
InChi Key: DCNUQRBLZWSGAV-UHFFFAOYSA-N
SMILES: O=CN(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results